missing translation for 'onlineSavingsMsg'
Learn More

CERILLIANT™ alpha-Linolenic acid, 1.0 mg/mL in Ethanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001546
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01001546 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01001546 Supplier Merck Emd Millipore Supplier No. L0391ML

Certified reference material

α-Linolenic acid, also known as ALA, is an omega-3 fatty acid essential for human health and well-being. Measurement of ALA levels in serum is useful for evaluating essential fatty acid defficiency (EFAD), malabsorption and ALA treatment response in patients with EFAD. This certified solution standard is suitable for use as starting material in calibrators and controls for GC/MS fatty acid testing methods in clinical and diagnostic testing of ALA in patient samples.

Specifications

CAS 64-17-5, 463-40-1
Linear Formula C18H30O2
MDL Number MFCD00065720
UN Number UN 1170 - class 3 - PG 2 - Ethanol
Concentration 1.0 mg/mL in ethanol
Formula Weight 278.43
Packaging Ampule
Quantity 1 mL
Flash Point 14°C (57.2°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation