Learn More
CERILLIANT™ ADB-FUBINACA Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
Certified reference material
Supplier: Merck Emd Millipore S1101ML
Description
ADB-FUBINACA is a potent synthetic cannabinoid receptor agonist that has been found in Spice-type herbal blends. This certified Snap-N-Spike™ solution is suitable as starting material for calibrators or controls in synthetic cannabinoid GC/MS or LC/MS methods for urine drug testing, forensic analysis, or clinical toxicology applications. For those synthetic cannabinoids currently controlled at the federal level, our certified solution standards of these substances are DEA-exempt, allowing laboratories to place orders without additional regulatory paperwork. For US customers, state specific regulatory requirements may apply.Specifications
| 67-56-1,1185282-00-1 | |
| 100 μg/mL in methanol | |
| Ampule | |
| 9.7°C (49.5°F) - closed cup |
| C21H23FN4O2 | |
| 382.43 | |
| 1 mL |
CERILLIANT is a registered trademark of Cerilliant Corporation