missing translation for 'onlineSavingsMsg'
Learn More

CERILLIANT™ Acryl fentanyl Hydrochloride Solution, 100 μg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™

Catalog No. 01001021
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01001021 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01001021 Supplier Merck Emd Millipore Supplier No. A1400.5ML

Certified reference material

Acryl fentanyl, also known as acryloylfentanyl, is a highly potent opioid analgesic fentanyl analog that is sold as designer drug and abused recreationally. A certified Snap-N-Spike™ solution suitable for use as starting material in calibrators and controls in acryl fentanyl LC/MS or GC/MS testing methods for clinical toxicology, urine drug testing or forensic analysis applications.

Specifications

Packaging Ampule
CAS 79279-03-1
Flash Point 9.7°C (49.5°F) - closed cup
Concentration 100 μg/mL in methanol (as free base)
Linear Formula C22H26N2O · HCl
MDL Number MFCD30718302
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Quantity 0.5 mL
Formula Weight 370.92

CERILLIANT is a registered trademark of Cerilliant Corporation