Learn More
CERILLIANT™ Acetyl norfentanyl-13C6 oxalate Solution, 100 μg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™
Certified reference material
Supplier: Merck Emd Millipore A1161ML
Description
An internal standard suitable for acetyl norfentanyl testing of urine or other matrices by GC/MS or LC/MS in applications such as pain prescription monitoring, clinical toxicology, urine drug testing, or forensic analysis. Acetyl norfentanyl is a major metabolite of acetyl fentanyl, a designer drug and fentanyl analog with a potency 40 times greater than heroin and 80 times greater than morphine. In June, 2013, the Centers for Disease Control (CDC) issued a health alert for acetyl fentanyl, reporting 14 overdose deaths in Rhode Island.Specifications
| Ampule | |
| 9.7°C (49.5°F) - closed cup | |
| C713C6H18N2O · C2H2O4 | |
| 1 mL |
| 67-56-1 | |
| 100 μg/mL in methanol (as free base) | |
| UN 1230 - class 3 - PG 2 - Methanol | |
| 314.29 |
CERILLIANT is a registered trademark of Cerilliant Corporation