missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CERILLIANT™ Acetyl fentanyl Solution, 50 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
Description
A certified Snap-N-Spike™ solution suitable for use as starting material in calibrators and controls for fentanyl LC/MS or GC/MS testing methods in clinical toxicology, urine drug testing, prescription monitoring, or forensic analysis applications. Acetyl fentanyl is a designer drug and fentanyl analog with a potency 40 times greater than heroin and 80 times greater than morphine. This Certified Spiking Solution™ is supplied in a convenient, quantitative, US DEA-exempt solution format and with TK No.'s for Canadian customers.
Specifications
Specifications
| Packaging | Ampule |
| CAS | 67-56-1 |
| Flash Point | 9.7°C (49.5°F) - closed cup |
| Concentration | 50 μg/mL in methanol |
| Linear Formula | C21H26N2O |
| MDL Number | MFCD07369982 |
| UN Number | UN 1230 - class 3 - PG 2 - Methanol, solution |
| Quantity | 1 mL |
| Formula Weight | 322.44 |
CERILLIANT is a registered trademark of Cerilliant Corporation