missing translation for 'onlineSavingsMsg'
Learn More

CERILLIANT™ Acetyl fentanyl Solution, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001004
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01001004 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01001004 Supplier Merck Emd Millipore Supplier No. A1091ML

Certified reference material

A certified Snap-N-Spike™ solution suitable for use as starting material in calibrators and controls for fentanyl LC/MS or GC/MS testing methods in clinical toxicology, urine drug testing, prescription monitoring, or forensic analysis applications. Acetyl fentanyl is a designer drug and fentanyl analog with a potency 40 times greater than heroin and 80 times greater than morphine. In June, 2013, the Centers for Disease Control (CDC) issued a health alert for acetyl fentanyl, reporting 14 overdose deaths in Rhode Island.

Specifications

Packaging Ampule
CAS 67-56-1
Concentration 1.0 mg/mL in methanol
Linear Formula C21H26N2O
MDL Number MFCD07369982
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Quantity 1 mL
Formula Weight 322.44

CERILLIANT is a registered trademark of Cerilliant Corporation