missing translation for 'onlineSavingsMsg'
Learn More

CERILLIANT™ Acetyl fentanyl-13C6 Solution, 50 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001017
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01001017 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01001017 Supplier Merck Emd Millipore Supplier No. A1301ML

Certified reference material

Acetyl fentanyl is a designer drug and fentanyl analog with a potency 40 times greater than heroin and 80 times greater than morphine. Acetyl fentanyl has been directly implicated in numerous opiate overdose deaths throughout the United States. This stable-labeled internal standard is suitable for quantification of acetyl fentanyl levels in urine or other matrices by LC/MS or GC/MS for clinical toxicology, forensic analysis, prescription monitoring, or urine drug testing applications. This Certified Spiking Solution™ is supplied in a convenient, quantitative, US DEA-exempt solution format and with TK No.'s for Canadian customers.

Specifications

Packaging Ampule
CAS 67-56-1
Flash Point 9.7°C (49.5°F) - closed cup
Concentration 50 μg/mL in methanol
Linear Formula 13C6C15H26N2O
MDL Number MFCD07369982
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Quantity 1 mL
Formula Weight 328.4

CERILLIANT is a registered trademark of Cerilliant Corporation