missing translation for 'onlineSavingsMsg'
Learn More

CERILLIANT™ Acetyl fentanyl-13C6 Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001005
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01001005 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01001005 Supplier Merck Emd Millipore Supplier No. A1101ML
Only null left

Certified reference material

Acetyl fentanyl is a designer drug and fentanyl analog with a potency 40 times greater than heroin and 80 times greater than morphine. In June, 2013, the Centers for Disease Control (CDC) issued a health alert for acetyl fentanyl, reporting 14 overdose deaths in Rhode Island. This stable-labeled internal standard is suitable for quantification of acetyl fentanyl levels in urine or other matrices by LC/MS or GC/MS for clinical toxicology, forensic analysis, prescription monitoring, or urine drug testing applications.

Specifications

Packaging Ampule
CAS 67-56-1
Flash Point 9.7°C (49.5°F)
Concentration 100 μg/mL in methanol
Linear Formula 13C6C15H26N2O
MDL Number MFCD07369982
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Quantity 1 mL
Formula Weight 328.4

CERILLIANT is a registered trademark of Cerilliant Corporation