missing translation for 'onlineSavingsMsg'
Learn More

CERILLIANT™ Acetaminophen-D4 Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001908
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01001908 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01001908 Supplier Merck Emd Millipore Supplier No. P9091ML

Certified reference material

A stable-labeled internal standard suitable for quantitation of acetaminophen levels in urine, serum, or plasma by LC/MS or GC/MS for clinical toxicology, isotope dilution methods, P450 studies, and other pharmaceutical research. Acetaminophen, or Paracetamol, is a common over-the-counter COX inhibitor drug used for pain relief and fever reduction.

Specifications

CAS 67-56-1, 64315-36-2
Linear Formula C8D4H5NO2
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 100 μg/mL in methanol
Formula Weight 155.19
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation