missing translation for 'onlineSavingsMsg'
Learn More

CERILLIANT™ AB-PINACA 5-Pentanoic acid metabolite, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001986
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01001986 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01001986 Supplier Merck Emd Millipore Supplier No. S0851ML
Only null left

Certified reference material

AB-PINACA 5-Pentanoic acid metabolite is a major urinary metabolite of the synthetic cannabinoid AB-PINACA. Over the past couple of years, clusters of cases of severe toxic effects and deaths related to AB-PINACA use have increased in the United States. This certified reference solution is suitable for use in synthetic cannabinoid testing methods by LC/MS or GC/MS for applications in clinical toxicology, urine drug testing, and forensic analysis.

Specifications

CAS 67-56-1, 1630022-93-3
Linear Formula C18H24N4O4
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 100 μg/mL in methanol
Formula Weight 360.41
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation