missing translation for 'onlineSavingsMsg'
Learn More

CERILLIANT™ 7-Carboxy cannabidiol (7-COOH-CBD) Solution, 1 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001132
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01001132 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01001132 Supplier Merck Emd Millipore Supplier No. C1811ML

Certified reference material

Specifications

CAS 67-56-1
Linear Formula C21H28O4
Concentration 1 mg/mL in methanol
Formula Weight 344.44
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation