Learn More
CERILLIANT™ 7-Aminonitrazepam-D5 Solution, 100 μg/mL in Acetonitrile, Sold by MilliporeSigma™ Supelco™
Certified reference material
Supplier: Merck Emd Millipore A1261ML
Description
7-Aminonitrazepam is a major urinary metabolite of nitrazepam, a benzodiazepine used to treat insomnia and sold under the trade names Mogadon and Alodorm. This internal standard is suitable for quantitation of 7-Aminonitrazepam levels using LC/MS or GC/MS methods for urine drug testing, clinical toxicology, or forensic analysis applications.Specifications
| 75-05-8 | |
| UN 1648 3 / PGII | |
| 256.31 | |
| 1 mL |
| C15H8D5N3O | |
| 100 μg/mL in acetonitrile | |
| Ampule | |
| 2°C (35.6°F) - closed cup |
CERILLIANT is a registered trademark of Cerilliant Corporation