missing translation for 'onlineSavingsMsg'
Learn More

CERILLIANT™ 5F-MDMB-PINACA (5F-ADB) Solution, 100 μg/mL in Acetonitrile, Sold by MilliporeSigma™ Supelco™

Catalog No. 01002009
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01002009 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01002009 Supplier Merck Emd Millipore Supplier No. S1131ML
Only null left

Certified reference material

5F-MDMB-PINACA, also known as 5F-ADB, is a synthetic cannabinoid used as an active ingredient in synthetic cannabis products and is sold online as a designer drug. A certified solution standard suitable for use in LC/MS or GC/MS synthetic cannabinoid testing applications including clinical toxicology, forensic analysis, and urine drug testing.

Specifications

CAS 75-05-8, 1838134-16-9
Linear Formula C20H28FN3O3
Concentration 100 μg/mL in acetonitrile
Formula Weight 377.45
Packaging Ampule
Quantity 1 mL
Flash Point 2°C (35.6°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation