missing translation for 'onlineSavingsMsg'
Learn More

CERILLIANT™ 5-MeO-DMT Solution, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001650
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01001650 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01001650 Supplier Merck Emd Millipore Supplier No. M1681ML

Certified reference material

5-MeO-DMT is a powerful, hallucinogenic tryptamine found in various plants used historically in the preparation of psychotropic snuffs. This certified solution standard is suitable for 5-MeO-DMT testing methods by GC/MS or LC/MS in clinical toxicology, urine drug testing, or forensic analysis applications.

Specifications

CAS 67-56-1, 1019-45-0
Linear Formula C13H18N2O
MDL Number MFCD00005658
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 1.0 mg/mL in methanol
Formula Weight 218.29
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation