missing translation for 'onlineSavingsMsg'
Learn More

CERILLIANT™ 5-Fluoro PB-22 3-carboxyindole metabolite Solution, 100 μg/mL in Acetonitrile, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001997
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01001997 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01001997 Supplier Merck Emd Millipore Supplier No. S1011ML

Certified reference material

5-Fluoro PB-22 3-carboxyindole is a major metabolite of the synthetic cannabinoid 5-Fluoro PB-22, also known as 5F-PB-22. This certified Snap-N-Spike™ solution is suitable as starting material for calibrators or controls in synthetic cannabinoid GC/MS or LC/MS methods for urine drug testing, forensic analysis, or clinical toxicology applications. For those synthetic cannabinoids currently controlled at the federal level, our certified solution standards of these substances are DEA-exempt, allowing laboratories to place orders without additional regulatory paperwork. For US customers, state specific regulatory requirements may apply.

Specifications

CAS 75-05-8, 1432794-98-3
Linear Formula C14H16FNO2
MDL Number MFCD28134508
UN Number UN 1648 3 / PGII
Concentration 100 μg/mL in acetonitrile
Formula Weight 249.28
Packaging Ampule
Quantity 1 mL
Flash Point 2°C (35.6°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation