missing translation for 'onlineSavingsMsg'
Learn More

CERILLIANT™ 4-Methylethcathinone Hydrochloride Solution, 1.0 mg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™

Catalog No. 01001642
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01001642 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01001642 Supplier Merck Emd Millipore Supplier No. M1551ML

Certified reference material

Also called 4-MEC, 4-Methylethcathinone is a stimulant and bath salt cathinone. The designer drug is often sold in mixtures with other substituted cathinones under the name 'NRG-2.' In addition to bath salt mixtures, some countries have even reported use of 4-MEC as the active ingredient in 'Ecstasy' pills.

Specifications

Packaging Ampule
CAS 67-56-1
Flash Point 9.7°C (49.5°F) - closed cup
Concentration 1.0 mg/mL in methanol (as free base)
Linear Formula C12H18ClNO
MDL Number MFCD19313132
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Quantity 1 mL
Formula Weight 227.73

CERILLIANT is a registered trademark of Cerilliant Corporation