missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CERILLIANT™ 4'-Hydroxydiclofenac Solution, 100 μg/mL in Acetonitrile, Sold by MilliporeSigma™ Supelco™
Description
A certified solution standard suitable for use in LC/MS or GC/MS applications for clinical toxicology, forensic analysis, or pharmaceutical research. 4′-Hydroxydiclofenac is a major metabolite of diclofenac, a COX-2 inhibitor used for the treatment of minor aches, pains, and fever.
Specifications
Specifications
| CAS | 75-05-8, 64118-84-9 |
| Linear Formula | C14H11Cl2NO3 |
| UN Number | UN 1648 3 / PGII |
| Concentration | 100 μg/mL in acetonitrile |
| Formula Weight | 312.15 |
| Packaging | Ampule |
| Quantity | 1 mL |
| Flash Point | 2°C (35.6°F) - closed cup |
CERILLIANT is a registered trademark of Cerilliant Corporation