missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CERILLIANT™ 4-Fluoromethcathinone Hydrochloride Solution, 1.0 mg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™
Description
4-Fluoromethcathinone is a stimulant and new designer drug found in 'bath salts' sold at head shops and over the Internet. These synthetic cathinones offer recreational uses that mimic the effects of illegal drugs such as cocaine, methamphetamine, and LSD. Abuse of 4-fluoromethcathinone, also called flephedrone or 4-FMC, has been observed throughout the US from Louisiana, Texas, and Florida to New Jersey, Illinois, and Kansas.
Specifications
Specifications
| Packaging | Ampule |
| CAS | 7589-35-7 |
| Flash Point | 9.7°C (49.5°F) - closed cup |
| Concentration | 1.0 mg/mL in methanol (as free base) |
| Linear Formula | C10H13ClFNO |
| MDL Number | MFCD22495221 |
| UN Number | UN 1230 - class 3 - PG 2 - Methanol, solution |
| Synonym | Flephedrone hydrochloride |
| Quantity | 1 mL |
| Formula Weight | 217.67 |
CERILLIANT is a registered trademark of Cerilliant Corporation