missing translation for 'onlineSavingsMsg'
Learn More

CERILLIANT™ 4-Fluoro-isobutyryl fentanyl Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001374
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01001374 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01001374 Supplier Merck Emd Millipore Supplier No. F0500.5ML

Certified reference material

4-Fluoro-isobutyryl fentanyl, also known as 4-FIBF and p-FIBF, is a fentanyl analog and sold on the illicit market as designer drug. This certified Snap-N-Spike™ solution is suitable for use as starting material in calibrators and controls for 4-Fluoro-isobutyryl fentanyl LC/MS or GC/MS testing methods in clinical toxicology, urine drug testing, or forensic analysis applications. This certified solution standard is DEA-exempt, allowing laboratories to place orders without additional regulatory paperwork. For US customers, state specific regulatory requirements may apply.

Specifications

Packaging Ampule
CAS 67-56-1
Flash Point 9.7°C (49.5°F) - closed cup
Concentration 100 μg/mL in methanol
Linear Formula C23H29FN2O
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Quantity 0.5 mL
Formula Weight 368.49

CERILLIANT is a registered trademark of Cerilliant Corporation