missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CERILLIANT™ 4-ANPP-D5 Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
Description
4-ANPP, also known as Despropionyl fentanyl, is an intermediate in the synthesis of fentanyl and related opioids and is often found as an impurity in fentanyl preparations. This stable-labeled internal standard is suitable for quantification of 4-ANPP levels in urine or other matrices by LC/MS or GC/MS for clinical toxicology, forensic analysis, prescription monitoring, or urine drug testing applications. This Certified Spiking Solution™ is supplied in a convenient US DEA-exempt solution format.
Specifications
Specifications
| Packaging | Ampule |
| CAS | 67-56-1 |
| Flash Point | 9.7°C (49.5°F) - closed cup |
| Concentration | 100 μg/mL in methanol |
| Linear Formula | C19H19D5N2 |
| UN Number | UN 1230 - class 3 - PG 2 - Methanol, solution |
| Quantity | 0.5 mL |
| Formula Weight | 285.44 |
CERILLIANT is a registered trademark of Cerilliant Corporation