missing translation for 'onlineSavingsMsg'
Learn More

CERILLIANT™ 3-Hydroxyphenazepam Solution, 1.0 mg/mL in Acetonitrile, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001461
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01001461 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01001461 Supplier Merck Emd Millipore Supplier No. H1131ML
Only null left

Certified reference material

3-Hydroxyphenazepam is an active metabolite of the novel psychoactive benzodiazepine phenazepam, which emerged in online drug markets in late 2011. Despite being a controlled substance in Europe, North America, and many countries in Asia, phenazepam has been encountered in the form of LSD mimic blotters and as counterfeit diazepam. This certified Snap-N-Spike™ solution is suitable for use as a starting material in calibrators and controls for numerous GC/MS or LC/MS applications in forensic analysis, clinical toxicology and urine drug testing.

Specifications

CAS 75-05-8, 70030-11-4
Linear Formula C15H10BrClN2O2
MDL Number MFCD00413715
UN Number UN 1648 3 / PGII
Concentration 1.0 mg/mL in acetonitrile
Formula Weight 365.61
Packaging Ampule
Quantity 1 mL
Flash Point 2°C (35.6°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation