missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CERILLIANT™ 3-epi-25-Hydroxyvitamin D3 Solution, 50 μg/mL in Ethanol, Sold by MilliporeSigma™ Supelco™
Description
A Certified Spiking Solution™ of 3-epi-25-Hydroxyvitamin D3, the epi isomer of the prohormone 25-hydroxyvitamin D3. The presence of 3-epi-25-hydroxyvitamin D3 can affect accurate quantitation of 25-hydroxyvitamin D3 levels by most routine LC-MS/MS methods and to an unknown extent in immuno- and protein binding assays. This solution standard is suitable for critical quantitative applications in research and clinical environments including: for use in preparation of controls and calibrators for LC/UV and LC-MS/MS assays; as system tuning/suitability standards; linearity verification; internal proficiency testing; and for OEM use in preparation of reagents, development of assays and instrument IQ/OQ.
Specifications
Specifications
| CAS | 64-17-5, 73809-05-9 |
| Melting Point | -144.0°C |
| Linear Formula | C27H44O2 |
| MDL Number | MFCD11656130 |
| UN Number | UN 1170 - class 3 - PG 2 - Ethanol |
| Concentration | 50 μg/mL in ethanol |
| Formula Weight | 400.64 |
| Packaging | Ampule |
| Quantity | 1 mL |
| Flash Point | 14°C (57.2°F) - closed cup |
CERILLIANT is a registered trademark of Cerilliant Corporation