missing translation for 'onlineSavingsMsg'
Learn More

CERILLIANT™ 3,4-Methylenedioxypyrovalerone HCl (MDPV) Solution, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001635
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01001635 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01001635 Supplier Merck Emd Millipore Supplier No. M1461ML

Certified reference material

3,4-Methylenedioxypyrovalerone, also known as MDPV, Mtv, or Super Coke, is a new designer drug and stimulant with cases of abuse reported in Europe and the US. In mid- to late-2010, MDPV started appearing in the US as a legal drug alternative marketed as “bath salts” under names including Ivory Wave, Tranquility, and Cloud 9. This certified solution standard offers numerous applications in GC/MS or LC/MS testing from forensic analysis and clinical toxicology to urine drug monitoring.

Specifications

Packaging Ampule
CAS 67-56-1
Flash Point 9.7°C (49.5°F) - closed cup
Concentration 1.0 mg/mL in methanol (as free base)
Linear Formula C16H21NO3 · HCl
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Synonym 3,4-Methylenedioxypyrovalerone hydrochloride, MDPV hydrochloride, MDPV
Quantity 1 mL
Formula Weight 311.8

CERILLIANT is a registered trademark of Cerilliant Corporation