missing translation for 'onlineSavingsMsg'
Learn More

CERILLIANT™ 25C-NBOMe-D3 Hydrochloride Solution, 100 μg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™

Catalog No. 01001093
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01001093 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01001093 Supplier Merck Emd Millipore Supplier No. C1321ML

Certified reference material

25C-NBOMe is a second generation 2C amine derived from attaching an NBOMe (2-methoxybenzyl) group to the amine group of 2C-C. Sold under street names such as N-Bomb, Boom, and C-Boom, this hallucinogenic phenethylamine is active at microgram doses. This internal standard is suitable for quantitation of 25C-NBOMe in LC/MS or GC/MS testing methods for forensic analysis, urine drug testing, or clinical toxicology applications.

Specifications

Packaging Ampule
CAS 67-56-1
Flash Point 9.7°C (49.5°F) - closed cup
Concentration 100 μg/mL in methanol (as free base)
Linear Formula C18H19D3ClNO3 · HCl
UN Number UN 1230 - class 3 - PG 2 - Methanol
Quantity 1 mL
Formula Weight 375.3

CERILLIANT is a registered trademark of Cerilliant Corporation