missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CERILLIANT™ 2,4,6-Trinitrotoluene Solution, 10 mg/mL in Acetonitrile, Sold by MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 75-05-8, 118-96-7 |
| Linear Formula | C7H5N3O6 |
| MDL Number | MFCD00041875 |
| UN Number | UN 1648 3 / PGII |
| Concentration | 10 mg/mL in acetonitrile |
| Synonym | TNT |
| Formula Weight | 227.13 |
| Packaging | Ampule |
| Quantity | 5 mL |
| Flash Point | 2°C (35.6°F) - closed cup |
CERILLIANT is a registered trademark of Cerilliant Corporation