missing translation for 'onlineSavingsMsg'
Learn More

CERILLIANT™ 1alpha,25-Dihydroxyvitamin D2 Solution, 5 μg/mL in Ethanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001443
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01001443 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01001443 Supplier Merck Emd Millipore Supplier No. H0901ML

Certified reference material

Accurate preparation of a quantitative reference is highly critical for accurate assessment of patient Vitamin D levels. This Certified Spiking Solution™ is suitable for critical quantitative applications in research and clinical environments including: for use in preparation of controls and calibrators for LC/UV and LC-MS/MS Vitamin D assays; as system tuning/suitability standards; linearity verification; internal proficiency testing; and for OEM use in preparation of reagents, development of assays and instrument IQ/OQ. The stable, quantitative solution format provides significant benefits to the user including material savings, increased efficiency of labor, and convenience of use. The shelf stable Snap-N-Spike™ format allows laboratories to eliminate painstaking weighing operations and provides the ability to spike a known concentration of analyte into their matrix of choice just before use.

Specifications

CAS 64-17-5, 60133-18-8
Linear Formula C28H44O3
UN Number UN 1170 - class 3 - PG 2 - Ethanol, solution
Concentration 5 μg/mL in ethanol
Formula Weight 428.65
Packaging Ampule
Quantity 1 mL
Flash Point 14°C (57.2°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation