missing translation for 'onlineSavingsMsg'
Learn More

CERILLIANT™ 17alpha-Methyltestosterone Solution, 1.0 mg/mL, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001693
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01001693 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01001693 Supplier Merck Emd Millipore Supplier No. M9061ML

Certified reference material

17α-Methyltestosterone is an androgenic steroid used in the treatment of testosterone defficiency. 17a-Methyltestosterone is abused in sports as a performance-enhancing drug. This analytical reference standard is applicable for use in sports testing, endocrinology, urine drug testing, or forensic analysis by LC-MS/MS or GC/MS.

Specifications

CAS 110-71-4, 58-18-4
Linear Formula C20H30O2
MDL Number MFCD00003655
UN Number UN 2252 - class 3 - PG 2 - 1,2-Dimethoxyethane solution
Concentration 1.0 mg/mL
Synonym 17alpha-Methyl-4-androsten-17beta-ol-3-one solution, 17beta-Hydroxy-17alpha-methyl-4-androsten-3-one solution, Mesterone solution, Methyltestosterone solution
Formula Weight 302.45
Packaging Ampule
Quantity 1 mL
Flash Point -2°C (28.4°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation