missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CERILLIANT™ (+/-)-11-nor-9-Carboxy-Delta9-THC-D3 Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
Description
11-nor-9-Carboxy-δ9-THC, also known as THC acid, is the main secondary metabolite of δ9-THC found in urine or blood of cannabis users. This internal standard is applicable for numerous applications in GC/MS or LC/MS testing from forensic analysis and clinical toxicology to urine drug testing and isotope dilution methods.
Specifications
Specifications
| CAS | 67-56-1, 136844-96-7 |
| Linear Formula | C21H25O4D3 |
| MDL Number | MFCD00673309 |
| UN Number | UN 1230 - class 3 - PG 2 - Methanol, solution |
| Concentration | 100 μg/mL in methanol |
| Formula Weight | 347.46 |
| Packaging | Ampule |
| Quantity | 1 mL |
| Flash Point | 9.7°C (49.5°F) - closed cup |
CERILLIANT is a registered trademark of Cerilliant Corporation