Learn More
CERILLIANT™ 1-(3-Chlorophenyl)piperazine Hydrochloride Solution, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
Certified reference material
Supplier: Merck Emd Millipore C0891ML
Description
1-(3-Chlorophenyl)-piperazine, also known as meta-chlorophenylpiperazine or mCPP, is a designer drug analog of aryl-substituted piperazines. A stimulant, serotonin antagonist and reuptake inhibitor (SARU), mCPP can also be detected as a primary urinary metabolite of the antidepressant drug trazadone. This certified solution standard is suitable for use in piperazine testing methods by LC/MS or GC/MS for clinical toxicology, forensic analysis, or urine drug testing applications.Specifications
| 67-56-1,65369-76-8 | |
| UN 1230 - class 3 - PG 2 - Methanol, solution | |
| 1-(3-Chlorophenyl)piperazine ammonium salt | |
| Ampule | |
| 9.7°C (49.5°F) - closed cup |
| C10H14Cl2N2 | |
| 1.0 mg/mL in methanol (as free base) | |
| 233.14 | |
| 1 mL |
CERILLIANT is a registered trademark of Cerilliant Corporation