missing translation for 'onlineSavingsMsg'
Learn More

Ceric Sulfate Solution, 0.1N (N/10) (Certified), Fisher Chemical

Catalog No. SC661
Change view
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
SC661 1 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SC661 Supplier Thermo Fisher Scientific Supplier No. SC66 1
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

EcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by an SFI certified supplier.

Chemical Identifiers

CAS 17106-39-7 , 7664-93-9 , 7732-18-5
Molecular Formula CeO8S2
Molecular Weight (g/mol) 332.23
MDL Number MFCD00148852
InChI Key VZDYWEUILIUIDF-UHFFFAOYSA-J
Synonym ceric sulfate, cerium iv sulfate, ceric disulfate, ceric sulphate, cerium disulfate, cerium 4+ sulfate, cerium 4+ disulphate, unii-4f66fi5t7x, sulfuric acid, cerium 4+ salt 2:1, cerium 4+ disulfate
PubChem CID 159684
IUPAC Name λ⁴-cerium(4+) disulfate
SMILES [Ce+4].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Specifications

Color Yellow
Physical Form Liquid
Quantity 1 L
Grade Certified
Concentration or Composition (by Analyte or Components) 0.1N (N/10)
Identification Pass Test
Packaging Glass Bottle
Vapor Pressure 14mmHg at 20°C
Density Vapor Density: 0.7g/mol
Chemical Name or Material Ceric Sulfate Solution, 0.1N (N/10)