missing translation for 'onlineSavingsMsg'
Learn More

Ceric Sulfate, Reagent, 85%, Spectrum™ Chemical

C1204, 13590-82-4, Ce(SO4)2

Supplier:  Spectrum Chemical Mfg Cor C1204500GM

 View more versions of this product

Catalog No. 18601542

Only null left
Explore available promotions

Description

Description

Spectrum™ Chemical Ceric Sulfate, Reagent is an inorganic compound that is a yellow solid that is water soluble and used as a potent oxidizer. It is used for redox titration in analytical chemistry. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.
Specifications

Specifications

13590-82-4
CeO8S2
500 g
[Ce+4].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
332.23
Reagent
Poly Bottle
100%
MFCD00148852
VZDYWEUILIUIDF-UHFFFAOYSA-J
λ⁴-cerium(4+) disulfate
85%
0.1%

Chemical Identifiers

13590-82-4
332.23
VZDYWEUILIUIDF-UHFFFAOYSA-J
[Ce+4].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
CeO8S2
MFCD00148852
λ⁴-cerium(4+) disulfate
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions