missing translation for 'onlineSavingsMsg'
Learn More

Ceric Sulfate, Reagent, 85%, Spectrum™ Chemical

Catalog No. p-7157136
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
20 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18601541 100 g Poly Bottle
18601542 500 g Poly Bottle
18601544 2.5 kg Poly Pail
18601543 20 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18601541 Supplier Spectrum Chemical Mfg Cor Supplier No. C1204100GM
Only null left

C1204, 13590-82-4, Ce(SO4)2

Spectrum™ Chemical Ceric Sulfate, Reagent is an inorganic compound that is a yellow solid that is water soluble and used as a potent oxidizer. It is used for redox titration in analytical chemistry. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.

Chemical Identifiers

CAS 13590-82-4
Molecular Formula CeO8S2
Molecular Weight (g/mol) 332.23
MDL Number MFCD00148852
InChI Key VZDYWEUILIUIDF-UHFFFAOYSA-J
IUPAC Name λ⁴-cerium(4+) disulfate
SMILES [Ce+4].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Specifications

CAS Max % 100%
Quantity 100 g
Percent Purity 85%
Grade Reagent
Insoluble Matter 0.1%
Packaging Poly Bottle