missing translation for 'onlineSavingsMsg'
Learn More

Ceric Sulfate, 0.100 Normal (N/10) In Dilute Sulfuric Acid, Reagents

Catalog No. 01336520
Change view
Click to view available options
Quantity:
20 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01336520 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01336520 Supplier Labchem Inc Supplier No. C717225020F

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ceric Sulfate, 0.100 Normal (N/10) In Dilute Sulfuric Acid, Reagent, for general laboratory use. Manufactured in ISO 9001 facility. NIST Traceable.

Chemical Identifiers

CAS 7732-18-5 , 7664-93-9 , 7664-93-9
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Cerium(IV) Sulfate
IUPAC Name water
SMILES O

Specifications

Boiling Point 100°C
Color Orange
pH <1
Physical Form Liquid
Quantity 20 L
Grade Reagent
Packaging Cubitainer
Specific Gravity 1.085
Chemical Name or Material Ceric Sulfate, 0.100 Normal (N/10) In Dilute Sulfuric Acid