missing translation for 'onlineSavingsMsg'
Learn More

Ceric Ammonium Sulfate, in 1.0N Sulfuric Acid, Certified, 0.100N ±0.001N (0.10M), LabChem™

Catalog No. p-5388305
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
Packaging:
Amber Glass
Cubitainer™
Catalog No. Quantity Packaging
LC129301 500 mL Amber Glass
LC129302 1 L Amber Glass
LC129304 4 L Amber Glass
LC129306 10 L Cubitainer™
LC129305 20 L Cubitainer™
5 options

Catalog No. LC129301

Supplier: LabChem LC129301

Only null left

Chemical Identifiers

CAS 16774-21-3 , 7732-18-5 , 7664-93-9
Molecular Formula CeH8N8O18
Molecular Weight (g/mol) 548.22
InChI Key WIBGOERAEYJBOT-UHFFFAOYSA-N
Synonym Ceric ammonium nitrate, CAN
PubChem CID 24181046  
IUPAC Name λ⁴-cerium(4+) bis(nitric acid) diamine tetranitrate
SMILES N.N.[Ce+4].O[N+]([O-])=O.O[N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Color Orange
Concentration 0.100N ±0.001N (0.10M)
CAS Max % 5.7, 91.3, 3
Identification Passes Test
Packaging Amber Glass
Quantity 500 mL
Physical Form Liquid
Linear Formula Ce(NH4)2(NO3)6
UN Number UN2796
Solubility Information Soluble in water
Formula Weight 548.23
Grade Certified
Concentration or Composition (by Analyte or Components) 0.100N ±0.001N (0.10M)
Chemical Name or Material Ceric Ammonium Sulfate, in 1.0N Sulfuric Acid
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature