Learn More
Ceric Ammonium Sulfate, in 1.0N Sulfuric Acid, Certified, 0.100N ±0.001N (0.10M), LabChem™
Supplier: LabChem LC129304
Specifications
| Ceric Ammonium Sulfate | |
| in 1.0N Sulfuric Acid | |
| 0.100N ±0.001N (0.10M) | |
| Passes Test | |
| 4 L | |
| Liquid | |
| UN2796 | |
| Soluble in water | |
| N.N.[Ce+4].O[N+]([O-])=O.O[N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O | |
| 548.22 | |
| 548.23 | |
| 0.100N ±0.001N (0.10M) |
| Orange | |
| 16774-21-3,7732-18-5,7664-93-9 | |
| 5.7,91.3,3 | |
| Amber Glass | |
| CeH8N8O18 | |
| Ce(NH4)2(NO3)6 | |
| Ceric ammonium nitrate, CAN | |
| WIBGOERAEYJBOT-UHFFFAOYSA-N | |
| λâ´-cerium(4+) bis(nitric acid) diamine tetranitrate | |
| 24181046 | |
| Certified |
Chemical Identifiers
| 16774-21-3 | |
| 548.22 | |
| Ceric ammonium nitrate, CAN | |
| λâ´-cerium(4+) bis(nitric acid) diamine tetranitrate |
| CeH8N8O18 | |
| WIBGOERAEYJBOT-UHFFFAOYSA-N | |
| 24181046 | |
| N.N.[Ce+4].O[N+]([O-])=O.O[N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Danger
Recommended Storage : Room Temperature