missing translation for 'onlineSavingsMsg'
Learn More

Ceric Ammonium Sulfate, in 1.0N Sulfuric Acid, Certified, 0.100N ±0.001N (0.10M), LabChem™

Supplier:  LabChem LC129302

 View more versions of this product

Catalog No. LC129302


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Ceric Ammonium Sulfate
in 1.0N Sulfuric Acid
0.100N ±0.001N (0.10M)
Passes Test
1 L
Liquid
UN2796
Soluble in water
N.N.[Ce+4].O[N+]([O-])=O.O[N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
548.22
548.23
0.100N ±0.001N (0.10M)
Orange
16774-21-3,7732-18-5,7664-93-9
5.7,91.3,3
Amber Glass
CeH8N8O18
Ce(NH4)2(NO3)6
Ceric ammonium nitrate, CAN
WIBGOERAEYJBOT-UHFFFAOYSA-N
λ⁴-cerium(4+) bis(nitric acid) diamine tetranitrate
24181046
Certified

Chemical Identifiers

16774-21-3
548.22
Ceric ammonium nitrate, CAN
λ⁴-cerium(4+) bis(nitric acid) diamine tetranitrate
CeH8N8O18
WIBGOERAEYJBOT-UHFFFAOYSA-N
24181046
N.N.[Ce+4].O[N+]([O-])=O.O[N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

Danger

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions