missing translation for 'onlineSavingsMsg'
Learn More

Ceric Ammonium Sulfate, 0.100 Normal in 1 N Sulfuric Acid, Reagents

Catalog No. 01335695
Change view
Click to view available options
Quantity:
1 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01335695 1 L
01335694 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01335695 Supplier Labchem Inc Supplier No. CC0076001A
Only null left

Ceric Ammonium Sulfate, 0.100 Normal in 1 N Sulfuric Acid, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 10378-47-9
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Ammonium Cerium(IV) Sulfate Dihydrate
IUPAC Name water
SMILES O

Specifications

Boiling Point 102°C
Color Orange
pH <1
Physical Form Liquid
Quantity 1 L
Grade Reagent
Packaging Poly Bottle
Specific Gravity 1.03
Chemical Name or Material Ceric Ammonium Sulfate, 0.100 Normal in 1 N Sulfuric Acid