missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cephradine Monohydrate 96.0+%, TCI America™
Specifications
Specifications
| Melting Point | 186°C |
| Color | White-Yellow |
| Quantity | 5 g |
| Formula Weight | 349.41 |
| Percent Purity | ≥96.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Cephradine Monohydrate |
Safety and Handling
| RTECSNumber | XID336000 |
| TSCA | No |
| Recommended Storage | Refrigerator |