accessibility menu, dialog, popup

UserName

CAS RN 75975-70-1

IUPAC Name: 7-[2-amino-2-(cyclohexa-1,4-dien-1-yl)acetamido]-3-methyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid hydrate
Synonyms: cephradine hydrate
Molecular Weight (g/mol): 367.42
Molecular Formula: C16H21N3O5S
InChi Key: VHNPSPMQGXQSET-UHFFFAOYNA-N
SMILES: O.CC1=C(N2C(SC1)C(NC(=O)C(N)C1=CCC=CC1)C2=O)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results