missing translation for 'onlineSavingsMsg'
Learn More

Cephradine Monohydrate 96.0+%, TCI America™

Catalog No. C29881G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C29881G 1 g
C29885G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C29881G Supplier TCI America Supplier No. C29881G

Chemical Identifiers

CAS 75975-70-1
Molecular Formula C16H21N3O5S
Molecular Weight (g/mol) 367.42
MDL Number MFCD00865048
InChI Key VHNPSPMQGXQSET-CYJZLJNKSA-N
PubChem CID 21124775
IUPAC Name (6R,7R)-7-[[(2R)-2-amino-2-cyclohexa-1,4-dien-1-ylacetyl]amino]-3-methyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid;hydrate
SMILES CC1=C(N2C(C(C2=O)NC(=O)C(C3=CCC=CC3)N)SC1)C(=O)O.O

Specifications

Melting Point 186°C
Color White-Yellow
Quantity 1 g
Formula Weight 349.41
Percent Purity ≥96.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Cephradine Monohydrate
RTECSNumber XID336000
TSCA No
Recommended Storage Refrigerator