missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Cephalothin | |
| 154°C | |
| C16H15N2NaO6S2 | |
| 25 g | |
| VUFGUVLLDPOSBC-XRZFDKQNSA-M | |
| sodium (6R,7R)-3-[(acetyloxy)methyl]-8-oxo-7-[2-(thiophen-2-yl)acetamido]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate | |
| 6024 | |
| 396.43 | |
| Crystalline Powder |
| 153-61-7 | |
| Yellow | |
| MFCD00242614 | |
| Cephalotin, 7-(2-Thienylacetamido)cephalosporanic Acid | |
| [Na+].CC(=O)OCC1=C(N2[C@H](SC1)[C@H](NC(=O)CC1=CC=CS1)C2=O)C([O-])=O | |
| 418.41 | |
| CHEBI:124991 | |
| ≥98.0% (HPLC,T) |
Chemical Identifiers
| 153-61-7 | |
| 418.41 | |
| VUFGUVLLDPOSBC-XRZFDKQNSA-M | |
| 6024 | |
| sodium (6R,7R)-3-[(acetyloxy)methyl]-8-oxo-7-[2-(thiophen-2-yl)acetamido]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate |
| C16H15N2NaO6S2 | |
| MFCD00242614 | |
| Cephalotin, 7-(2-Thienylacetamido)cephalosporanic Acid | |
| CHEBI:124991 | |
| [Na+].CC(=O)OCC1=C(N2[C@H](SC1)[C@H](NC(=O)CC1=CC=CS1)C2=O)C([O-])=O |
Safety and Handling
RTECSNumber : XI0388000
TSCA : No