missing translation for 'onlineSavingsMsg'
Learn More

Cephalothin 98.0+%, TCI America™

Catalog No. p-7125644
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C27695G 5 g
C276925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C27695G Supplier TCI America Supplier No. C27695G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 153-61-7
Molecular Formula C16H15N2NaO6S2
Molecular Weight (g/mol) 418.41
MDL Number MFCD00242614
InChI Key VUFGUVLLDPOSBC-XRZFDKQNSA-M
Synonym Cephalotin, 7-(2-Thienylacetamido)cephalosporanic Acid
PubChem CID 6024
ChEBI CHEBI:124991
IUPAC Name sodium (6R,7R)-3-[(acetyloxy)methyl]-8-oxo-7-[2-(thiophen-2-yl)acetamido]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
SMILES [Na+].CC(=O)OCC1=C(N2[C@H](SC1)[C@H](NC(=O)CC1=CC=CS1)C2=O)C([O-])=O

Specifications

Melting Point 154°C
Color Yellow
Quantity 5 g
Formula Weight 396.43
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Cephalothin
RTECSNumber XI0388000
TSCA No