missing translation for 'onlineSavingsMsg'
Learn More

Cephalexin Monohydrate USP MP Biomedicals

Catalog No. p-3178716
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15058505 5 g
ICN15058525 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15058505 Supplier MP Biomedicals Inc Supplier No. 0215058505
Only null left

Chemical Identifiers

CAS 23325-78-2
Molecular Formula C16H19N3O5S
Molecular Weight (g/mol) 365.40
MDL Number MFCD00167148,MFCD00056877
InChI Key AVGYWQBCYZHHPN-CYJZLJNKSA-N
Synonym 7-(d-a-Aminophenylacetamido)desacetoxycephalosporanic acid, Cefalexin monohydrate
PubChem CID 62921
ChEBI CHEBI:3535
IUPAC Name (6R,7R)-7-[(2R)-2-amino-2-phenylacetamido]-3-methyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid hydrate
SMILES O.CC1=C(N2[C@H](SC1)[C@H](NC(=O)[C@H](N)C1=CC=CC=C1)C2=O)C(O)=O

Specifications

Type Cephalexin Monohydrate
Physical Form Crystalline Powder
Color Beige to White
Quantity 5 g
Formula Weight 365.4
Grade BP/USP
Assay ≥95%
Chemical Name or Material Cephalexin Monohydrate
EINECSNumber 239-773-6
Recommended Storage Store at 4°C, desiccate.