missing translation for 'onlineSavingsMsg'
Learn More

Cephalexin hydrate, 97+%

Catalog No. AAJ6317206
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6317206 5 g
AAJ6317214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6317206 Supplier Thermo Scientific Chemicals Supplier No. J6317206
Only null left

Cephalexin hydrate, cefalexin hydrate, CAS # 23325-78-2, is a first-generation cephalosporin antibiotic from the beta-lactam family. | CAS: 15686-71-2 | C16H20ClN3O5S | 401.862 g/mol

Cephalexin is a cephalosporin antibiotic used to study the effect of expression, binding, and inhibition of PBP3 and other penicillin-binding proteins (PBPs) on bacterial cell wall mucopeptide synthesis. Cephalexin disrupts the synthesis of the peptidoglycan layer of bacterial cell walls which is responsible for cell wall structural integrity. Peptidoglycan synthesis is facilitated by transpeptidases known as penicillin-binding proteins (PBPs). PBPs bind to the D-Ala-D-Ala at the end of muropeptides (peptidoglycan precursors) to crosslink the peptidoglycan. Cephalexin antibiotics mimic the D-Ala-D-Ala site, thereby competitively inhibiting PBP crosslinking of peptidoglycan.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Cephalexin hydrate is the hydrate of the cephalosporin antibiotic, cephalexin

• Cephalexin is a beta-lactam antibiotic that inhibits bacterial growth and division

• Cephalexin binds to penicillin-binding protein and inhibits cell wall synthesis

Application

• Cephalexin works best on gram-positive microbes

• Useful for immunology research, particularly on antibiotic allergies and cross-reactivity

Chemical Identifiers

CAS 15686-71-2
Molecular Formula C16H20ClN3O5S
Molecular Weight (g/mol) 401.862
MDL Number MFCD00167148
InChI Key YHJDZIQOCSDIQU-OEDJVVDHSA-N
PubChem CID 57515946
IUPAC Name (6R,7R)-7-[[(2R)-2-amino-2-phenylacetyl]amino]-3-methyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid;hydron;chloride;hydrate
SMILES [H+].CC1=C(N2C(C(C2=O)NC(=O)C(C3=CC=CC=C3)N)SC1)C(=O)O.O.[Cl-]

Specifications

Type Cephalexin Hydrate
Physical Form Powder
Color White
Quantity 5 g
Beilstein 965503
Merck Index 141974
Solubility Information Soluble in DMSO and Methanol.
Formula Weight 347.39 (Anhydrous)
Percent Purity ≥97%
Chemical Name or Material Cephalexin hydrate
Hazard Category Respiratory sensitisation (category 1), Skin sensitisation (category 1)
Hazard Statement May cause allergy or asthma symptoms or breathing difficulties if inhaled, May cause an allergic skin reaction
EINECSNumber 239-773-6
RTECSNumber XI0350000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only