missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CEP55 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA529454
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA529454 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA529454 Supplier Invitrogen™ Supplier No. PA529454
Only null left

Rabbit Polyclonal Antibody

Recommended positive controls: IMR32, U87-MG, MCF-7, HeLa. Predicted reactivity: Rat (80%), Bovine (87%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Plays a role in mitotic exit and cytokinesis (PubMed:16198290, PubMed:17853893). Recruits PDCD6IP and TSG101 to midbody during cytokinesis. Required for successful completion of cytokinesis (PubMed:17853893). Not required for microtubule nucleation (PubMed:16198290). Plays a role in the development of the brain and kidney (PubMed:28264986). [UniProt].

Specifications

Antigen CEP55
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.85 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene CEP55
Gene Accession No. Q53EZ4
Gene Alias 1200008O12Rik; 2700032M20Rik; C10orf3; cancer/testis antigen 111; centrosomal protein 55; centrosomal protein 55kDa; centrosomal protein of 55 kDa; centrosomal protein of 55 kDa; LOW QUALITY PROTEIN: centrosomal protein of 55 kDa; centrosomal protein of 55 kDa-like protein; CEP55; CT111; RGD1305340; Up-regulated in colon cancer 6; URCC6
Gene Symbols CEP55
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human CEP55. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55165
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less