missing translation for 'onlineSavingsMsg'
Learn More

CEP131/AZ1 Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501560333
Encompass
Change view
Click to view available options
Quantity:
10 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501560333 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 501560333 Supplier Bethyl Laboratories Inc Supplier No. A301416AT

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

CEP131 (Centrosomal Protein 131) is a Protein Coding gene. Diseases associated with CEP131 includeAnterior Spinal Artery Syndrome. Among its related pathways areOrganelle biogenesis and mainteceandCell Cycle, Mitotic. Gene Ontology (GO) annotations related to this gene includeprotein homodimerization activity.

Specifications

Antigen CEP131/AZ1
Applications Immunoprecipitation
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene CEP131
Gene Accession No. Q9UPN4
Gene Alias 5-azacytidine induced 1, 5-azacytidine-induced protein 1, AZ1, AZI1, centrosomal protein 131 kDa, centrosomal protein 131kDa, KIAA1118, pre-acrosome localization protein 1, ZA1
Gene Symbols CEP131
Host Species Rabbit
Immunogen Between 325 and 375
Purification Method Antigen affinity chromatography
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22994
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less