Learn More
CEP Oil Only Pads and Rolls
Contain and adsorb oils while repelling aquatic liquids
Supplier: CEP P200
Description
- Constructed of meltblown polypropylene fibers for cost-effective adsorption
- Fast-wicking fibers absorb up to 25 times their weight in hydrocarbon
- Choice of pads and rolls with varying sorbing capacities
Specifications
| 25.2 gal. | |
| Polypropylene Fiber | |
| Fast-wicking fibers absorb up to 25 times their weight in hydrocarbon | |
| 17 x 15 in. | |
| 43.18 cm | |
| 38 cm |
| 95.3 L | |
| Sorbent Pad | |
| Single-weight | |
| 17 in. | |
| 15 in. |