Learn More
CEP Oil Only Pads and Rolls
Contain and adsorb oils while repelling aquatic liquids
Supplier:  CEP FBESR144
 View more versions of this product
Catalog No. 19500529
Only null left
Explore available promotions
Description
Description
  • Constructed of meltblown polypropylene fibers for cost-effective adsorption
  • Fast-wicking fibers absorb up to 25 times their weight in hydrocarbon
  • Choice of pads and rolls with varying sorbing capacities
Specifications
Specifications
39 gal.
Polypropylene Fiber
Fast-wicking fibers absorb up to 25 times their weight in hydrocarbon
150 ft. x 15 in.
45.7 m
38 cm
147.6 L
Sorbent Roll
Medium-weight
150 ft.
15 in.
Product Suggestions
Product Suggestions