missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Centrifuge/Stirrer

Catalog No. 14127570
Change view
Click to view available options
Capacity:
12 x 1.5/2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity
14127570 12 x 1.5/2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14127570 Supplier Fisherbrand™ Supplier No. 120583

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The Fisherbrand™ Centrifuge / Stirrer is the industry's first high-speed centrifuge (12,500 rpm) and magnetic stirrer (3 L), combined into one unit. This design innovation enables a substantial cost savings for labs by not having to purchase two pieces of equipment, a stirrer and a centrifuge, because the Fisherbrand™ Centrifuge / Stirrer does it all.

5 Year Warranty

Fisherbrand™ Centrifuge / Stirrer:

  • Made from a durable chemically resistant ABS and polycarbonate making it easy to clean, maintenance free and helps lower the cost of ownership.
  • The modern low-profile design is compact and has a small footprint for labs with limited bench space and limited height that ultimately helps reduced clutter in the lab.
  • An easy to use back-lit digital control panel for time, speed and operations ensures effective outcomes and streamlines the work flow process by allowing for quick set up.
  • Capacity: Centrifuge: 12 place 1.5/2.0 mL tube rotor: 24 mL total maximum liquid volume, 6 place 5 mL tube rotor: 30 mL total maximum liquid volume, Stirrer: 3 liters plus.
  • Diameter of vessel not to exceed 130 mm (6.70 in.).
  • Includes: 5 mL Tube Rotor
  • 1.5/2.0 mL Tube Rotor
  • Magnetic Stirrer Rotor
  • 12 Tube adapters 0.2 mL
  • 12 Tube adapters for 0.5 mL
  • 2 Silicone mats for stirrer lid
  • 2 Stir Bars
  • 1 low voltage, double insulated power adapter with 4 interchangeable plugs

High-Speed Mini Centrifuge:

  • The high-speed Centrifuge (12,500 rpm) is designed to give you maximum speed and flexibility while being comfortable and easy to use.
  • Ideal for most protocols requiring fast spins (12,500 rpm / 9,800 x g), and appropriate for PCR, microfilter cell separation, and HPLC protocols.
  • Adjustable time (30 seconds to 60 minutes) and speed (500 to 12,500 rpm) - Variable speed can be set in RPM or RCF and has rapid acceleration and fast controlled breaking.
  • Two rotors included, 12 place for 1.5/2.0 mL tubes and 6 place for 5 mL tubes with adapters for additional tube sizes.
  • Manual lid override allows users to open lid in case of a power failure and prevents loss of samples.
  • Safety features help ensure it will not operate when the lid is open, a rotor is jammed, or the unit is out of balance.
  • Compatible Tubes: - 0.2 mL, 0.5 mL, 1.5 mL, 2.0 mL, 5.0 mL

Magnetic Stirrer:

  • The high-performance Magnetic Stirrer (3L Plus) is designed to provide a powerful magnetic coupling that ensures consistent accurate alignment of the stir bar that significantly reduce the chance of spin-outs.
  • Ideal for work with sensitive samples such as cell lines and protocols requiring more aggressive mixing to quickly achieve full dilution of samples in solution.
  • Multiple stirring modes, clockwise, c. clockwise and oscillate enable protocols to be controlled precisely for reproducible results.
  • Max. Speed: 12,500 rpm Centrifuge, 2,500 rpm Stirrer
  • Adjustable time (30 seconds to 60 minutes or continuous) and speed (50 to 2,500 RPM).
  • After hours of continuous use, the unit remains cool ensuring sample integrity.
  • Sensor technology prevents the stirrer rotor from being used above 2,500 rpm helping to protect users and prevent unit damage.
  • A silicone mat is included to help prevent spills and to help keep vessels stable.
  • High-quality rare-earth magnets are used in the stirrer rotor to ensure the strongest magnetic coupling.
  • Two rare-earth stir bars (40 x 12 mm) included have PTFE coating for chemical resistance.

Certifications:

CE, SGS, RoHS2, WEEE

  • EN/IEC 61010-1 ed. 3.1, 2017
  • EN/IEC 61010-2-051 ed.3, 2015
  • UL Std. 61010-1 ed.3, 2012
  • UL Std. 61010-2-051 ed.3, 2015
  • CAN/CSA C22.2 No.61010-

1-12 (R2017)

  • CAN/CSA C22.2 No.61010-

2-051 (R2015)

  • IEC 61326-1:2013 (IEC 61326-1:2012)
  • FCC Part 15, Subpart B: 2015
  • ICES-003, Issue 6

Warranty

5 Years

Specifications

Capacity 12 x 1.5/2 mL
Max. Speed 12,500 rpm (Centrifuge), 2,500 rpm (Stirrer)
Max. RCF 9,783 x g
Timer Yes
Certifications/Compliance CE, SGS, RoHS2, WEEE
Amperage 6.5 A
Safety Features Imbalance Safety cut-out, Tilt Safety cut-out, Safety Interlock Lid
Electrical Requirements Universal 12 V DC input
Dimensions (L x W x H) Exterior 9.2 x 7.3 x 4.7 in., Stirrer plate: Approx. 130 mm diameter
Controller Type Rotary Encoder
Rotor Included Yes
Warranty 5 Years
Refrigerated No
Run Time Centrifuge: 30 seconds to 60 minutes (5 second increments), Stirrer: Stirrer Mode 30 seconds to 60 minutes or continuous (5 second increments), Oscillating Stirrer Mode 1 second to 60 minutes or continuous, Oscillating Period 30 seconds to 15 minutes
Net Weight 4.08 lb.
Program Storage Last Run Memory
Frequency 50/60 Hz
Voltage 100/240 V
Rotor 5 mL Tube Rotor, 1.5/2.0 mL Tube Rotor, Magnetic Stirrer Rotor
Product Type Centrifuge/Stirrer
Display Type Digital
Show More Show Less