Learn More
Eppendorf™ Centrifuge 5910 Ri - Refrigerated Centrifuge
The refrigerated Eppendorf Centrifuge 5910 Ri stands out with an intuitive touch screen interface and a range of innovative solutions for usability, documentation and user management. Its wide variety of fixed-angle and swing-bucket rotors accommodate a comprehensive spectrum of applications in a compact footprint.
Supplier:  Eppendorf™ 5943000114
 View more versions of this product
Catalog No. 05414458
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Alert:
Centrifuge 5910 Ri is exclusively intended for indoor use. All country-specific safety requirements for operating electrical equipment in the lab must be observed.
Description
Description
  • Large 7-inch VisioNize™ touch interface for super-fast parameter settings via unique favorites function
  • User management with multiple authorization levels and documentation features supporting GLP/GxP requirements
  • Aerosol-tight Eppendorf QuickLock™ caps and lids for fast and ergonomic lid locking
  • Quiet operation to improve your work environment
  • Centrifuge lid with soft-touch lid closure
  • Automatic rotor recognition and imbalance detection for maximum operational safety
  • Swing-bucket rotors and adapters for tubes and bottles from 0.2 mL to 1,000 mL as well as all types of MTP, PCR or Deepwell Plates
  • Fixed-angle rotors for high-speed molecular biology applications in tubes from 0.2 mL to 250 mL
Specifications
Chemical Identifiers
VisioNize™ Touch Interface
Yes
10
-11°C to +40°C
72 x 68 x 37 cm (28.3 x 25.9 x 14.5 in.)
85 cm
120 V, 50/60 Hz (US)
12 A
Eppendorf 5910 Ri
VisioNize™ Touch Interface
99 Programs
10 sec. to 99 hr. 59 min.
33.5 in.
109 kg
120 V
50/60 Hz
Specifications
VisioNize™ Touch Interface
4,347 x g (w/ incl. rotor)
Eppendorf 5910 Ri
VisioNize™ Touch Interface
22,132 x g
<53 db(A)
99 Programs
Yes
10 sec. to 99 hr. 59 min.
72 x 68 x 37 cm (28.3 x 25.9 x 14.5 in.)
85 cm
120 V, 50/60 Hz (US)
1650 W
50/60 Hz
4 x 1,000 mL (w/ inc. rotor)
4,500 rpm (w/ incl. rotor)
Yes
4 x Eppendorf Wide-neck Bottles (1 L) and Bottle Adapters
14,000 rpm
10
S-4xUniversal
-11°C to +40°C
5910 Ri, Big Volume Harvesting Solution
33.5 in.
109 kg
120 V
12 A
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
Centrifuge 5910 Ri is used for the separation of aqueous solutions and suspensions of different densities in approved sample tubes.